missing translation for 'onlineSavingsMsg'
Learn More

Oxazole 98.0+%, TCI America™

Catalog No. O02871G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O02871G Supplier TCI America Supplier No. O02871G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 288-42-6
Molecular Formula C3H3NO
Molecular Weight (g/mol) 69.063
MDL Number MFCD00009751
InChI Key ZCQWOFVYLHDMMC-UHFFFAOYSA-N
Synonym oxazole, unii-fjz20i1lps, fjz20i1lps, oxazol, 1,3-oxazol, pubchem8626, oxazole 1g
PubChem CID 9255
ChEBI CHEBI:35597
IUPAC Name 1,3-oxazole
SMILES C1=COC=N1

Specifications

Melting Point -82°C
Color Colorless
Boiling Point 70°C
Quantity 1 g
UN Number 1993
Formula Weight 69.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Oxazole