accessibility menu, dialog, popup

UserName

ON

CAS RN 288-42-6

IUPAC Name: 1,3-oxazole
Synonyms: oxazole
Molecular Weight (g/mol): 69.06
Molecular Formula: C3H3NO
InChi Key: ZCQWOFVYLHDMMC-UHFFFAOYSA-N
SMILES: O1C=CN=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)