missing translation for 'onlineSavingsMsg'
Learn More

Oxazole, 98+%

Catalog No. AAL1834103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1834103 1 g
AAL1834106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1834103 Supplier Thermo Scientific Chemicals Supplier No. L1834103

CAS: 288-42-6 | C3H3NO | 69.063 g/mol

Oxazole and its derivatives are actively involved as building blocks for bio chemicals and pharmaceuticals like flopristin and darglitazone. It is also involved in industrial applications such as in dyes, fluorescent brightening agents, textile auxiliaries and plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxazole and its derivatives are actively involved as building blocks for bio chemicals and pharmaceuticals like flopristin and darglitazone. It is also involved in industrial applications such as in dyes, fluorescent brightening agents, textile auxiliaries and plastics.

Solubility
Miscible with alcohol and ether. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 288-42-6
Molecular Formula C3H3NO
Molecular Weight (g/mol) 69.063
MDL Number MFCD00009751
InChI Key ZCQWOFVYLHDMMC-UHFFFAOYSA-N
Synonym oxazole, unii-fjz20i1lps, fjz20i1lps, oxazol, 1,3-oxazol, pubchem8626, oxazole 1g
PubChem CID 9255
ChEBI CHEBI:35597
IUPAC Name 1,3-oxazole
SMILES C1=COC=N1

Specifications

Melting Point -86°C
Density 1.05
Boiling Point 69°C to 70°C
Flash Point 19°C (66°F)
Refractive Index 1.427
Quantity 1 g
UN Number UN2924
Beilstein 103851
Solubility Information Miscible with alcohol and ether. Slightly miscible with water.
Formula Weight 69.07
Percent Purity ≥98%
Chemical Name or Material Oxazole
Show More Show Less
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 206-020-8
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only