missing translation for 'onlineSavingsMsg'
Learn More

Oxamic Acid 97.0+%, TCI America™

Catalog No. O00845G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O00845G 5 g
O008425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O00845G Supplier TCI America Supplier No. O00845G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 471-47-6
Molecular Formula C2H3NO3
Molecular Weight (g/mol) 89.05
MDL Number MFCD00008006
InChI Key SOWBFZRMHSNYGE-UHFFFAOYSA-N
Synonym 2-amino-2-oxoacetic acid, oxamate, acetic acid, aminooxo, oxalamic acid, oxamidic acid, glycine, 2-oxo, oxalic acid monoamide, amino oxo acetic acid, aminooxoacetic acid, oxamate repellent
PubChem CID 974
ChEBI CHEBI:18058
IUPAC Name oxamic acid
SMILES C(=O)(C(=O)O)N

Specifications

Melting Point 217°C
Color White
Quantity 5 g
Formula Weight 89.05
Percent Purity ≥97.0% (T,N)
Physical Form Crystalline Powder
Chemical Name or Material Oxamic Acid