missing translation for 'onlineSavingsMsg'
Learn More

Oxamic Acid Potassium Salt 98.0+%, TCI America™

Catalog No. p-7134220
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O016925G 25 g
O0169500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O016925G Supplier TCI America Supplier No. O016925G

Chemical Identifiers

CAS 21141-31-1
Molecular Formula C2H2KNO3
Molecular Weight (g/mol) 127.14
MDL Number MFCD00058475
InChI Key PSLCBKISHMXGLT-UHFFFAOYSA-M
Synonym Oxamidic Acid Potassium Salt, Potassium Oxamate
PubChem CID 23663672
IUPAC Name potassium carbamoylformate
SMILES [K+].NC(=O)C([O-])=O

Specifications

Color White
Quantity 25 g
Formula Weight 127.14
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Oxamic Acid Potassium Salt