accessibility menu, dialog, popup

UserName

CAS RN 21141-31-1

IUPAC Name: potassium carbamoylformate
Synonyms: potassium oxamate
Molecular Weight (g/mol): 127.14
Molecular Formula: C2H2KNO3
InChi Key: PSLCBKISHMXGLT-UHFFFAOYSA-M
SMILES: [K+].NC(=O)C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results