missing translation for 'onlineSavingsMsg'
Learn More

Oxamic acid, 98%

Catalog No. AAB2034406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2034406 5 g
AAB2034414 25 g
AAB2034422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2034406 Supplier Thermo Scientific Chemicals Supplier No. B2034406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 471-47-6 | C2H3NO3 | 89.05 g/mol

Oxamic acid could also inhibit lactic dehydrogenase in the intact Ehrlich ascites tumor cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxamic acid could also inhibit lactic dehydrogenase in the intact Ehrlich ascites tumor cell.

Solubility
Soluble in water 108 mg/mL.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 471-47-6
Molecular Formula C2H3NO3
Molecular Weight (g/mol) 89.05
MDL Number MFCD00008006
InChI Key SOWBFZRMHSNYGE-UHFFFAOYSA-N
Synonym 2-amino-2-oxoacetic acid, oxamate, acetic acid, aminooxo, oxalamic acid, oxamidic acid, glycine, 2-oxo, oxalic acid monoamide, amino oxo acetic acid, aminooxoacetic acid, oxamate repellent
PubChem CID 974
ChEBI CHEBI:18058
IUPAC Name oxamic acid
SMILES C(=O)(C(=O)O)N

Specifications

Melting Point ∼205°C (decomposition)
Quantity 5 g
Beilstein 1743294
Merck Index 14,6917
Solubility Information Soluble in water 108mg/mL.
Formula Weight 89.05
Percent Purity 98%
Chemical Name or Material Oxamic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-443-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only