missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalic acid solution, Volumetric, 0.5Â M (COOH)2 (1N), Honeywell Fluka™
Volumetric, 0.5Â M (COOH)2 (1N)
$406.08 - $406.08
Chemical Identifiers
| CAS | 144-62-7 |
|---|---|
| Molecular Formula | C2H2O4 |
| Molecular Weight (g/mol) | 90.034 |
| InChI Key | MUBZPKHOEPUJKR-UHFFFAOYSA-N |
| Synonym | ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico |
| PubChem CID | 971 |
| ChEBI | CHEBI:16995 |
| IUPAC Name | oxalic acid |
| SMILES | C(=O)(C(=O)O)O |
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
| 144-62-7 | |
| 90.034 | |
| ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico | |
| CHEBI:16995 | |
| C(=O)(C(=O)O)O |
| C2H2O4 | |
| MUBZPKHOEPUJKR-UHFFFAOYSA-N | |
| 971 | |
| oxalic acid |
Specifications
| 144-62-7 | |
| HOOCCOOH | |
| NONH for all modes of transport | |
| MUBZPKHOEPUJKR-UHFFFAOYSA-N | |
| oxalic acid | |
| 971 | |
| 90.03g/mol | |
| Oxalic acid solution |
| C2H2O4 | |
| MFCD00002573 | |
| ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico | |
| C(=O)(C(=O)O)O | |
| 90.034 | |
| CHEBI:16995 | |
| 1 L |
Safety and Handling
P280-P305 + P351 + P338