missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid , For Silica, Certified, 10.0% (w/v) ±0.5%, LabChem™

Catalog No. LC180802
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC180802 1 L
LC180801 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC180802 Supplier LabChem Supplier No. LC180802

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6153-56-6 , 7732-18-5
Molecular Formula C2H6O6
Molecular Weight (g/mol) 126.064
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
PubChem CID 61373
IUPAC Name oxalic acid;dihydrate
SMILES C(=O)(C(=O)O)O.O.O

Specifications

Color Colorless
CAS Max % 90, 10
Quantity 1 L
Packaging Poly Bottle
Physical Form Liquid
Linear Formula HO2CCO2H
Traceability to NIST Traceable to NIST
Solubility Information Soluble in water
Formula Weight 90.04
Percent Purity 10.0% ±0.5% (w/v)
Grade Certified
Chemical Name or Material Oxalic Acid, For Silica
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/at
Recommended Storage Room Temperature