missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid , For Silica, Certified, 10.0% (w/v) ±0.5%, LabChem™

Catalog No. LC180801
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC180801 500 mL
LC180802 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. LC180801 missing translation for 'mfr' LabChem missing translation for 'supplierNo' LC180801

Chemical Identifiers

CAS 6153-56-6 , 7732-18-5
Molecular Formula C2H6O6
Molecular Weight (g/mol) 126.064
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
PubChem CID 61373
IUPAC Name oxalic acid;dihydrate
SMILES C(=O)(C(=O)O)O.O.O

Specifications

Color Colorless
CAS Max % 90, 10
Linear Formula HO2CCO2H
Solubility Information Soluble in water
Formula Weight 90.04
Percent Purity 10.0% ±0.5% (w/v)
Grade Certified
Quantity 500 mL
Packaging Poly Bottle
Physical Form Liquid
Traceability to NIST Traceable to NIST
Chemical Name or Material Oxalic Acid, For Silica
Show More Show Less
missing translation for 'healthHazard1' Warning
missing translation for 'healthHazard2' GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
missing translation for 'healthHazard3' GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature