missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalic Acid Dihydrate, Reagents
Oxalic Acid Dihydrate, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C211050050F4
Specifications
| Oxalic Acid Dihydrate | |
| 6153-56-6 | |
| C2H6O6, 2H2O | |
| 162.1 | |
| 50 kg | |
| Solid |
| 101°C to 102°C | |
| Colorless/White | |
| Ethanedioic Acid dihydrate | |
| Pure | |
| Fiber Drum | |
| 1.653 |