missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid Dihydrate, Reagents

Catalog No. p-200020986
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337580 100 g
01337578 3 kg
01337577 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337580 Supplier Labchem Inc Supplier No. C2110500100C2
Only null left

Oxalic Acid Dihydrate, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Oxalic Acid Dihydrate
CAS 6153-56-6
Melting Point 101°C to 102°C
Color Colorless/White
Molecular Formula C2H6O6, 2H2O
Synonym Ethanedioic Acid dihydrate
Molecular Weight (g/mol) 162.1
Grade Pure
Quantity 100 g
Packaging Glass Bottle
Physical Form Solid
Specific Gravity 1.653
Show More Show Less