missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid, 5% (w/v), Ricca Chemical

Catalog No. R54655004A
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R54655004A 4 L Natural Poly Bottle
5465516 500 mL Natural Poly Bottle
5465532 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R54655004A Supplier Ricca Chemical Company Supplier No. R54655004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5 , 6153-56-6
Molecular Formula C2H6O6
Molecular Weight (g/mol) Mixture
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
PubChem CID 61373
IUPAC Name oxalic acid;dihydrate
SMILES C(=O)(C(=O)O)O.O.O

Specifications

Color Colorless
Quantity 4 L
Packaging Natural Poly Bottle
Physical Form Liquid
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
Percent Purity 5% w/v
Grade Laboratory
Chemical Name or Material Oxalic Acid, 5% (w/v) Aqueous Solution