missing translation for 'onlineSavingsMsg'
Learn More

Oxalic acid, 5% w/v aq. soln.

Catalog No. AA35619K2
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35619K2 1 L
AA35619K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA35619K2 Supplier Thermo Scientific Chemicals Supplier No. 035619K2

CAS: 144-62-7 | C2H2O4 | 90.034 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 144-62-7
Molecular Formula C2H2O4
Molecular Weight (g/mol) 90.034
MDL Number MFCD00002573
InChI Key MUBZPKHOEPUJKR-UHFFFAOYSA-N
Synonym ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico
PubChem CID 971
ChEBI CHEBI:16995
IUPAC Name oxalic acid
SMILES C(=O)(C(=O)O)O

Specifications

UN Number UN3265
Concentration 5% w/v aq. soln.
Formula Weight 90.04
Quantity 1 L
Odor Odorless
Physical Form Liquid
Chemical Name or Material Oxalic acid
Hazard Category H290-H318
Hazard Statement GHS H Statement
H318-H302-H312
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P234-P280-P305+P351+P338-P310-P390
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 205-634-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only