missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalic Acid, 10% w/v, Reagents
Specifications
Specifications
| Boiling Point | 101°C |
| Color | Colorless |
| Percent Purity | 10% w/v |
| Grade | Reagent |
| Quantity | 1 L |
| Packaging | Poly Bottle |
| Physical Form | Liquid |
| Specific Gravity | 1.0476 |
| Chemical Name or Material | Oxalic Acid, 10% w/v |