missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid, 10% w/v, Reagents

Catalog No. 01336554
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336554 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336554 Supplier Labchem Inc Supplier No. C71101501A
Only null left

Oxalic Acid, 10% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 6153-56-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Ethanedioic Acid
IUPAC Name water
SMILES O

Specifications

Boiling Point 101°C
Color Colorless
Percent Purity 10% w/v
Grade Reagent
Quantity 1 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.0476
Chemical Name or Material Oxalic Acid, 10% w/v