missing translation for 'onlineSavingsMsg'
Learn More

Oxalate, Ion chromatography standard solution, Specpure™, C2O4-2 1000μg/mL

Catalog No. AA42568AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42568AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA42568AE Supplier Thermo Scientific Chemicals Supplier No. 042568AE

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

MDL Number MFCD00012465

Specifications

Odor Odorless
Physical Form Liquid
Quantity 100 mL
Grade Specpure
Chemical Name or Material Oxalate, Ion chromatography standard solution, C2O4-2 1000μg/mL
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only