Learn More
Oxacillin sodium salt monohydrate, 815^mg/mg
CAS: 7240-38-2 | C19H23N3NaO6S | 444.458 g/mol
Supplier: Thermo Scientific Chemicals J6638003
Description
Used primarily against Gram-positive organisms namely resistant Staphylococcus species
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Used primarily against gram positive organisms namely resistant Staphylococcus species
Specifications
| Oxacillin Sodium Salt Monohydrate | |
| 815μg/mg | |
| 7240-38-2 | |
| White | |
| >110°C (230°F) | |
| MFCD00167146 | |
| 4287093 | |
| Freely soluble in water | |
| [HH].CC1=C(C(=NO1)C2=CC=CC=C2)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O.O.[Na] | |
| 444.458 | |
| 441.43 |
| Oxacillin sodium salt monohydrate | |
| Powder | |
| 188°C | |
| 1 g | |
| C19H23N3NaO6S | |
| 687°C | |
| 14,6904 | |
| VKNIYYBMVNGVMD-MBGFYOAHSA-N | |
| (2S,5R,6R)-3,3-dimethyl-6-[(5-methyl-3-phenyl-1,2-oxazole-4-carbonyl)amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium;hydrate | |
| 131673944 |
Chemical Identifiers
| 7240-38-2 | |
| 444.458 | |
| VKNIYYBMVNGVMD-MBGFYOAHSA-N | |
| (2S,5R,6R)-3,3-dimethyl-6-[(5-methyl-3-phenyl-1,2-oxazole-4-carbonyl)amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium;hydrate |
| C19H23N3NaO6S | |
| MFCD00167146 | |
| 131673944 | |
| [HH].CC1=C(C(=NO1)C2=CC=CC=C2)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O.O.[Na] |
Safety and Handling
P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
H315-H317-H319-H334-H335
EINECSNumber : 214-636-3
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only