missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ P82 Dual-Gel Electrophoresis Double Sided Vertical System, 8-10 x 10cm gel, 150-300mL buffer volume

Catalog No. P82
Change view
Click to view available options
Length (Metric) Gel:
8 to 10 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Gel
P82 8 to 10 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P82 Supplier Thermo Scientific™ Supplier No. P82

Produce flat, even banding and crisp resolution with the easy-to-use Thermo Scientific™ Owl™ P82 Dual-Gel Electrophoresis System.

Features:
  • Runs two 10 x 10cm gels or one gel when used with blocking plate; accommodates 8 x 10cm gels utilizing the provided adapter
  • Allows gels to easily be placed into the device
  • Wedge placed in front of the cassettes provides even pressure against the leakproof gasket, places the cassettes in proper running position
  • Two sizes of wedges accommodate varying thickness of precast gels
  • Flexible—use precast polyacrylamide gels or hand cast gels
  • No cooling required

Includes:

  • Inner buffer chambers
  • Buffer tank
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • Two wedges (1.27cm thick)
  • Two wedges (1.59cm thick)
  • Adapter plates for 10 × 8 gels (2)
  • Blocking plate

Specifications

Certifications/Compliance CE marked
Description Double Sided Vertical System, 8-10 x 10cm gel system, 150mL to 300mL buffer volume
Max. Current 15 mA
Product Line Owl
Type Dual-Gel Electrophoresis Double Sided Vertical System
Volume (Metric) Lower Buffer Chamber Max. 300 mL
Volume (Metric) Lower Buffer Chamber Min. 150 mL
Volume (Metric) Upper Buffer Chamber 150 mL
Dimensions (L x W x H) 5.88 x 4.75 x 6.69 in. (15.2 x 12.4 x 17 cm)
Length (English) 5.88 in.
Length (Metric) 15.2 cm
Length (Metric) Gel 8 to 10 cm
Width (English) 4.75 in.
Width (Metric) 12.4 cm
Width (Metric) Gel 10 cm
Height (English) 6.69 in.
Height (Metric) 17 cm
Max. Voltage 330 V
Show More Show Less
Are Bio-Rad power supplies compatible with Owl Electrophoresis systems?

The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.