Learn More
Thermo Scientific™ Owl™ Leveling Platform
Accommodate most applications with this three-point leveling platform which ensures a flat casting and running surface.
$328.55 - $328.55
| Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
|
09-528-177
|
Thermo Scientific™
BLP |
Leveling Platform |
Each for $328.55
|
|
|||||
Description
Accommodate most applications with the Thermo Scientific™ Owl™ Leveling Platform, a three-point leveling platform that ensures a flat casting and running surface.
- Size: 36 x 46cm
- Includes bubble level
Specifications
| Owl System | |
| Owl |