missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ JGC Gel Casting Systems
Description
Cast polyacrylamide gels faster with Thermo Scientific™ Owl™ JGC Gel Casting Systems, vertical protein gel casters that offer a unique casting method.
- Pouch collects excess polyacrylamide and seals bottom of gel, eliminating need for tape or bottom spacers.
- Design allows storage of gels without dehydration by adding buffer and heat sealing.
- System casts from one to four gels.
- Few moving parts result in outstanding durability, extra long life.
Includes:
- Casting stand
- Blocking plate
- Three-point leveling base
- Spacer placer
- 25 gel pouches
Specifications
Specifications
| For Use With (Application) | Gel Electrophoresis |
| For Use With (Equipment) | Owl JGC Gel Casting System |
| Gel Size | 20 x 20 cm |
| Product Line | Owl |
| Type | Gel Caster |
| Length (Metric) Gel | 14 cm |
| Width (English) | 7.06 in. |
| Width (Metric) | 18 cm |
| Width (Metric) Gel | 16 cm |
| Depth (English) | 6.69 in. |
| Show More |