missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ JGC Gel Casting Systems

Catalog No. OWJGC2
Change view
Click to view available options
Length (Metric) Gel:
14 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Gel
OWJGC2 14 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OWJGC2 Supplier Thermo Scientific™ Supplier No. JGC 2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cast polyacrylamide gels faster with Thermo Scientific™ Owl™ JGC Gel Casting Systems, vertical protein gel casters that offer a unique casting method.

Cast polyacrylamide gels faster with Thermo Scientific™ Owl™ JGC Gel Casting Systems, vertical protein gel casters that offer a unique casting method.

  • Pouch collects excess polyacrylamide and seals bottom of gel, eliminating need for tape or bottom spacers.
  • Design allows storage of gels without dehydration by adding buffer and heat sealing.
  • System casts from one to four gels.
  • Few moving parts result in outstanding durability, extra long life.

Includes:

  • Casting stand
  • Blocking plate
  • Three-point leveling base
  • Spacer placer
  • 25 gel pouches

Specifications

For Use With (Application) Gel Electrophoresis
For Use With (Equipment) Owl JGC Gel Casting System
Gel Size 20 x 20 cm
Product Line Owl
Type Gel Caster
Length (Metric) Gel 14 cm
Width (English) 7.06 in.
Width (Metric) 18 cm
Width (Metric) Gel 16 cm
Depth (English) 6.69 in.
Depth (Metric) 17 cm
Show More Show Less