missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems
Achieve superior flexibility in a small footprint with our B2 mini gel electrophoresis systems.
$1,512.03
Specifications
| Certifications/Compliance | CE marked |
|---|---|
| Gel Size | Mini |
| Product Line | Owl |
| Type | Mini Gel Electrophoresis System |
| Sample Size | 8 to 48 |
Description
Achieve superior flexibility in a small footprint with Thermo Scientific™ Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems. The simple-to-use casting method, as well as multiple comb and tray options, make this product Owl's most popular system.
Features:
- Two comb slots on the U.V. Transmissible (UVT) gel tray double sample capacity by doubling the number of sample wells
- UVT gel trays are silk screened with a florescent ruler for easy measurement of bands
Ordering Information:
Available with optional buffer exchange ports (B2-BP) for use with an external recirculation pump
Includes:
- Buffer chamber
- Thermo Scientific™ SuperSafe™ lid with attached power supply leads
- EasyCast gasketed U.V. Transmissible (UVT) gel tray
- Two combs: 12 & 20 well, double-sided, 1.0/1.5 mm thick
Specifications
| CE marked | |
| Owl | |
| 8 to 48 | |
| 9.63 x 7.06 x 3.75 in. (24.5 x 18 x 9.5 cm) | |
| 24.5 cm | |
| 7.06 in. | |
| 12 cm | |
| 9.5 cm |
| Mini | |
| Mini Gel Electrophoresis System | |
| 800 mL | |
| 9.63 in. | |
| 14 cm | |
| 18 cm | |
| 3.75 in. |