missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems

Catalog No. 09528118
Change view
Click to view available options
Includes:
EasyCast, Buffer Exchange Ports
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
09528118 EasyCast, Buffer Exchange Ports
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09528118 Supplier Thermo Scientific™ Supplier No. B2 BP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Achieve superior flexibility in a small footprint with our B2 mini gel electrophoresis systems.

Achieve superior flexibility in a small footprint with Thermo Scientific™ Owl™ EasyCast™ B2 Mini Gel Electrophoresis Systems. The simple-to-use casting method, as well as multiple comb and tray options, make this product Owl's most popular system.

  • Two comb slots on the U.V. Transmissible (UVT) gel tray double sample capacity by doubling the number of sample wells
  • UVT gel trays are silk screened with a florescent ruler for easy measurement of bands

Ordering Information:

Available with optional buffer exchange ports (B2-BP) for use with an external recirculation pump

Includes:

  • Buffer chamber
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • EasyCast gasketed U.V. Transmissible (UVT) gel tray
  • Two combs: 12 & 20 well, double-sided, 1.0/1.5 mm thick

Specifications

Certifications/Compliance CE marked
Gel Size Mini
Product Line Owl
Type Mini Gel Electrophoresis System
Includes EasyCast, Buffer Exchange Ports
Sample Size 8 to 48
Volume (Metric) Buffer 800 mL
Dimensions (L x W x H) 9.63 x 7.06 x 3.75 in. (24.5 x 18 x 9.5 cm)
Length (English) 9.63 in.
Length (Metric) 24.5 cm
Length (Metric) Gel 14 cm
Width (English) 7.06 in.
Width (Metric) 18 cm
Width (Metric) Gel 12 cm
Height (English) 3.75 in.
Height (Metric) 9.5 cm
Show More Show Less