missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems

Catalog No. OWP8DS3
Change view
Click to view available options
Volume (Metric) Buffer:
300 mL
Includes:
Apparatus Only
Complete System
Complete System plus Caster
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Buffer Includes
OWP8DS3 300 mL Apparatus Only
09518157B 300 mL Complete System plus Caster
OWP8DS2 300 mL Complete System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. OWP8DS3 Supplier Thermo Scientific™ Supplier No. P8DS3

Quickly and easily cast and run one or two gels in the same device with these Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems.

Quickly and easily cast and run one or two gels in the same device using Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems—comprised of the P8DS, P9DS and P10DS Dual Gel Systems.

Simple, Six-Step Use:

  1. Secure the glass cassettes
  2. Pour the agarose
  3. Plug in the specially designed trough
  4. Cast polyacrylamide solution in the cassettes
  5. Load samples
  6. Run the gel

Features:

  • Manufactured using a time-tested acrylic fabrication technology, providing a durable, long-lasting system
  • Central cooling core provides even cooling across the gel surface for outstanding resolution and clear banding
  • Cooling is achieved by circulating fluid through the core with external circulator or cool tap water
  • Very simple clamping mechanism makes loading glass cassettes and most manufacturers' precast gels trouble free

Recommended for:

  • SDS electrophoresis
  • The second dimension of two-dimensional (2-D) electrophoresis
  • Agarose electrophoresis
  • Native electrophoresis

Specifications

Product Line Owl
Type Dual-Gel Vertical Electrophoresis System Apparatus
Gel Dimensions 10 x 8-10 cm
Includes Apparatus Only
Model P8DS
Volume (Metric) Buffer 300 mL
Length (English) 6.7 in.
Length (Metric) Gel 8 to 10 cm
Width (English) 5.1 in.
Width (Metric) Gel 10 cm
Height (English) 5.5 in.
Show More Show Less