Learn More
Thermo Scientific™ Owl™ D3-14 System Replacement Parts
Description
Choose from various Thermo Scientific™ Owl™ D3-14 System Replacement Parts, including UVT gel tray, gaskets and external caster. For use with the Owl D3-14 System.
- UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
- Comb slots are positioned to enable up to 12 sample sets on the same gel
Specifications
Specifications
| Description | External caster (no trays/combs) |
| For Use With (Equipment) | Owl™ D3 Series Horizontal Electrophoresis System |
| Product Line | Owl |
| Type | Caster |