Learn More
Thermo Scientific™ Owl™ D3-14 System Replacement Parts
Catalog No. OWD3CST14
Change view
Click to view available options
Type:
Caster
Gasket
Gel Tray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
OWD3CST14 Caster
OWD3GK Gasket
09528164B Gel Tray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. OWD3CST14 Supplier Thermo Scientific™ Supplier No. D3CST14
Only null left
Choose from various Thermo Scientific™ Owl™ D3-14 System Replacement Parts including UVT gel tray, gaskets and external caster.
Choose from various Thermo Scientific™ Owl™ D3-14 System Replacement Parts, including UVT gel tray, gaskets and external caster. For use with the Owl D3-14 System.
  • UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
  • Comb slots are positioned to enable up to 12 sample sets on the same gel
Specifications
Description External caster (no trays/combs)
For Use With (Equipment) Owl™ D3 Series Horizontal Electrophoresis System
Product Line Owl
Type Caster