Learn More
Thermo Scientific™ A2 and A5 Large Gel System Replacement End Gates
Catalog No. OWA2EG
Change view
Click to view available options
Type:
End Gates
Catalog No. Type
OWA2EG End Gates
1 options
Catalog No. OWA2EG Supplier Thermo Scientific™ Supplier No. A2EG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Use these gasketed UVT gel trays, replacement gaskets and end gates with Thermo Scientific™ A2 and A5 Large Gel Systems.
Use Thermo Scientific™ A2 and A5 Large Gel System Replacement Parts with Owl A2 and A5 Large Gel Systems.
  • UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
  • Comb slots are positioned to allow 2, 3 or 4 sample sets to run equal lengths on the same gel
Specifications
For Use With (Equipment) Owl A2 and A5 Large Gel Electrophoresis System
Type End Gates
Quantity 1 Pair