missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Ovation Mechanical (M) Pipette

Catalog No. 09035046
Change view
Click to view available options
Volume (Metric):
2 μL
10 μL
20 μL
100 μL
200 μL
1000 μL
For Use With (Equipment):
10 μL, 20 μL Pipette Tips
1000 μL, 1200 μL Pipette Tips
200 μL Pipette Tips
300 μL Pipette Tips
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) For Use With (Equipment)
09035046 1000 μL 10 μL, 20 μL Pipette Tips
09035047 200 μL 200 μL Pipette Tips
09035048 200 μL 300 μL Pipette Tips
09035049 100 μL 300 μL Pipette Tips
09035050 100 μL 1000 μL, 1200 μL Pipette Tips
09035051 20 μL 10 μL, 20 μL Pipette Tips
09035052 10 μL 200 μL Pipette Tips
09035053 2 μL 200 μL Pipette Tips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 09035046 Supplier VistaLab™ Supplier No. 10701000LF

Ovation™ pipettes have a unique and patented ergonomic design. They are the only pipettes that fully address the issues of posture, repetition, and force to significantly reduce strain and pain that occur from pipetting, while always providing superior accuracy and precision.

  • The angled shape allows users to pipette with low arm and elbow height, decreasing tension and associated pain in the neck, shoulder, back, and elbow.
  • When used properly, the Ovation™ pipette will keep the user's hand and wrist in the neutral position recommended by ergonomic experts.
  • Contoured body shape and adjustable hook assures a comfortable, easy grip, and custom fit hand hold.
  • Spring-loaded tip acquisition means a gentle push is all that is required to attach tips, and an audible click confirms secure attachment.
  • Tips flick off easily with a low force push of the eject button.
  • Special filters in the nozzle prevent liquid from being inadvertently aspirated into the pipettor, protecting it from contamination.
  • Ovation™ pipettes are the only pipettes that stand on their own, which means tips never touch the bench, even if the pipette is put down in the middle of a procedure.
  • Mechanical models feature two-stroke operation, available with click stop or quick set digital volume adjustment (0.2 μL to 1000 μL)
  • Electronic models feature custom volume presets, multi-dispensing, mixing, and serial diluting (0.5 μL to 1250 μL).
  • Electronic Macro 5 mL and 10 mL, and electronic 8 and 12 multichannel models available.
  • Single channel models include a built-in micro tube cap opener.
  • Color-coded for easy and quick volume identification.

Specifications

Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Purple
Disposable Reusable
For Use With Pipette Tips
Grip Type Ergonomic
Material PVDF
Number of Channels 1
Programmability No
Sterility Non-sterile
Type Mechanical Pipette
Volume (Metric) 1000 μL
For Use With (Equipment) 10 μL, 20 μL Pipette Tips
Show More Show Less