missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies Ovation M

Catalog No. 10101121
Change view
Click to view available options
Volume (Metric):
0.2 to 2 μL
0.2 to 2 μL, 2 to 20 μL, 20 to 200 μL, 100 to 1000 μL
1 to 10 μL
1 to 10 μL, 10 to 100 μL, 100 to 1000 μL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
10101121 20 to 200 μL
10101117 0.2 to 2 μL
10101118 1 to 10 μL
10101119 2 to 20 μL
10101120 10 to 100 μL
10101124 100 to 1000 μL
10101122 1 to 10 μL, 10 to 100 μL, 100 to 1000 μL
10101123 0.2 to 2 μL, 2 to 20 μL, 20 to 200 μL, 100 to 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 10101121 Supplier VistaLab™ Technologies Supplier No. 10700200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Ovation M is the ideal ergonomic, manual pipette that is autoclavable for easy sterilization.

  • Mechanical 2-stroke (overblow) low-force plunger operation
  • Mechanical precise click stop volume adjustment
  • Autoclavable for easy sterilization
  • Built-in microtube cap opener
  • Shelf hanger included with each pipette
  • Two-year warranty

Specifications

Accuracy ±0.8% at 200 μL, ±0.8% at 10 μL, ±2.5% (at 20 μL)
Autoclavable Autoclavable
Color Blue
Disposable Reusable
For Use With Pipette Tips
Grip Type Ergonomic
Material Polycarbonate ABS
Number of Channels 1
Product Line Ovation M
Programmability No
Sterility Non-sterile
Type Manual Pipette
Volume (Metric) 20 to 200 μL
Includes Microtube Cap Opener
Increments 0.2 μL
Precision ±0.15% at 200 μL, ±0.25% at 100 μL, ±1% (at 20 μL)
Quantity 1 Ea.
Warranty 2 Years
Length (English) 10 in.
Width (English) 7 in.
Height (English) 3 in.
Show More Show Less