Learn More
VistaLab™ Technologies Ovation™ Electronic Single Channel Pipette PROMO
Catalog No. 22146101TI
Change view
Click to view available options
Accuracy:
±0.8%
Increments:
0.5 μL
Catalog No. Accuracy Increments
22146101TI ±0.8% 0.5 μL
1 options
Catalog No. 22146101TI Supplier VistaLab™ Technologies Supplier No. 10650125TI
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ideal ergonomic choice for those needing a comprehensive menu of automated liquid handling functions.
  • One-way disposable nozzle
  • Proprietary energy-release system discards tips at the push of a button
  • Contoured plunger electronically activates pipetting activity
  • Adjustable hook can be easily positioned for a custom fit and reduced holding effort
  • Bionatural shape gives user maximum stability when pipetting, with minimal contact pressure
  • Easy push-button controls
  • Fully electronic operations maximize efficiency for all tasks, including multiple dispensing, pipetting, reverse pipetting, mixing, serial diluting, and automatic sequential dispensing
  • Stands securely on countertops
These electronic single channel Ovation models require a power supply unit 22-147-115 (sold separately) to recharge the pipettes battery.
Electronic single channel models require an Ovation Power Supply unit (Mfr. No. 9060-9001; sold separately) to recharge the pipetter's battery
Specifications
Accuracy ±0.8%
Battery Type Lithium-ion
Compatible Tips VistaLab™ tip
For Use With VistaLab™ Tip
Model Right Handed
Number of Channels 1
Product Line Ovation
Type Pipette
Volume (Metric) 2 to 125 μL
Display LCD
Finger Style Ergonomic
For Use With (Equipment) VistaLab™ Tip
Increments 0.5 μL
Number of Speeds 5
Warranty 1 Year
Show More Show Less