missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Ovation Electronic Multichannel (E8) Pipette

Catalog No. 09035040
Change view
Click to view available options
Volume (Metric):
20 μL
125 μL
250 μL
1250 μL
Compatible Tips:
LfTS 10μL XL
LfTS 1200μL
LfTS 200μL
LfTS 300μL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Compatible Tips
09035040 1250 μL LfTS 1200μL
09035041 250 μL LfTS 300μL
09035042 250 μL LfTS 200μL
09035043 125 μL LfTS 300μL
09035044 125 μL LfTS 200μL
09035045 20 μL LfTS 10μL XL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 09035040 Supplier VistaLab™ Supplier No. 11601250LF

Ovation™ pipettes have a unique and patented ergonomic design. They are the only pipettes that fully address the issues of posture, repetition, and force to significantly reduce strain and pain that occur from pipetting, while always providing superior accuracy and precision.

  • The angled shape allows users to pipette with low arm and elbow height, decreasing tension and associated pain in the neck, shoulder, back, and elbow.
  • When used properly, the Ovation™ pipette will keep the user's hand and wrist in the neutral position recommended by ergonomic experts.
  • Contoured body shape and adjustable hook assures a comfortable, easy grip, and custom fit hand hold.
  • Spring-loaded tip acquisition means a gentle push is all that is required to attach tips, and an audible click confirms secure attachment.
  • Tips flick off easily with a low force push of the eject button.
  • Special filters in the nozzle prevent liquid from being inadvertently aspirated into the pipettor, protecting it from contamination.
  • Ovation™ pipettes are the only pipettes that stand on their own, which means tips never touch the bench, even if the pipette is put down in the middle of a procedure.
  • Mechanical models feature two-stroke operation, available with click stop or quick set digital volume adjustment (0.2 μL to 1000 μL)
  • Electronic models feature custom volume presets, multi-dispensing, mixing, and serial diluting (0.5 μL to 1250 μL).
  • Electronic Macro 5 mL and 10 mL, and electronic 8 and 12 multichannel models available.
  • Single channel models include a built-in micro tube cap opener.
  • Color-coded for easy and quick volume identification.

Specifications

Compatible Tips LfTS 1200μL
Adjustable Tip Spacing No
Autoclavable Not Autoclavable
Color Purple
Dispensing Speed Variable
Disposable Reusable
For Use With Pipette Tips
Grip Type Ergonomic
Material PVDF
Number of Channels 8
Programmability No
Sterility Non-sterile
Type Electronic Micropipette
Volume (Metric) 1250 μL
For Use With (Equipment) 300 μL Pipette Tips
Show More Show Less