missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OVA257-264 (SIINFEKL) peptide bound to H-2Kb Monoclonal Antibody (eBio25-D1.16 (25-D1.16)), Brilliant Ultra Violet™ 737, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 367574382
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
367574382 100 μg
367574380 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 367574382 Supplier Invitrogen™ Supplier No. 367574382
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

OVA257-264 (SIINFEKL) peptide bound to H-2Kb Monoclonal antibody specifically detects OVA257-264 (SIINFEKL) peptide bound to H-2Kb in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen OVA257-264 (SIINFEKL) peptide bound to H-2Kb
Applications Flow Cytometry
Classification Monoclonal
Clone eBio25-D1.16 (25-D1.16)
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 737
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Alias H-2Kb-SIINFEKL; OVA257-264; OVA257-264 H-2Kb; OVA-Kb
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less