Learn More
Cytiva Outlet Valve
Description
For use with the AKTA start automated protein purification system. The outlet valve is used to direct the flow to the fraction collector or to the waste.
Specifications
Specifications
| For Use With (Application) | Direct the flow to the fraction collector or to the waste |
| For Use With (Equipment) | AKTA start Automated Protein Purification System |