missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Ouchterlony Plates

Catalog No. ICN642771
Change view
Click to view available options
For Use With (Application):
Specific and low affinity antisera
Type:
Ouchterlony Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Type
ICN642771 Specific and low affinity antisera Ouchterlony Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN642771 Supplier MP Biomedicals™ Supplier No. 08642771

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for specific but low affinity antisera that requires properly spaced wells and olyethylene glycol in the agarose for optimal immuno -precipitation.

Specifications

Capacity (Metric) 10 μL
Material Polystyrene
Type Ouchterlony Plate
Well Count 7
For Use With (Application) Specific and low affinity antisera
Includes Agarose 1.2% and PEG 5%