missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osteopontin Monoclonal Antibody (K52025_9H8), Invitrogen™
Mouse Monoclonal Antibody
$1601.06 - $2821.92
Specifications
| Antigen | Osteopontin |
|---|---|
| Clone | K52025_9H8 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: DDQSAETHSH KQSRLYKRKA NDESNEHSDV IDSQELSKVS REFHSHEFHS HEDMLVVDPK SKEEDKHLKF
Osteopontin Monoclonal antibody specifically detects Osteopontin in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| Osteopontin | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 2AR; 2b7; 44 kDa bone phosphoprotein; Apl-1; BNSP; Bone sialoprotein 1; Bone sialoprotein 1 (BSPI/BSP1); Bsp; BSPI; Calcium oxalate crystal growth inhibitor protein; CALPHA1 fusion; Early T lymphocyte activation 1 (ETA1); early T-lymphocyte activation 1; early T-lymphocyte activation 1 protein; Eta; ETA-1; HGNC:11255; immunoglobulin alpha 1 heavy chain constant region fusion protein; MGC110940; Minopontin; nephropontin; Op; Opn; Opnl; OSP; osteopontin; osteopontin/immunoglobulin alpha 1 heavy chain constant region fusion protein; osteopontin-like protein; PSEC0156; Ric; Secreted phosphoprotein 1; secreted phosphoprotein 1 (osteopontin bone sialoprotein I early T lymphocyte activation 1); secreted phosphoprotein 1 (osteopontin, bone sialoprotein I, early T-lymphocyte activation 1); Secreted phosphoprotein 1 (SPP1); secreted phosphoprotein 1 variant 6; Sialoprotein (osteopontin); SPP1; SPP-1; SPP1/CALPHA1 fusion; unnamed protein product; urinar; urinary stone protein; Uropontin | |
| Recombinant human SPP1 (immunogen range is 231-300). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| SPP1 |
| K52025_9H8 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P10451 | |
| 6696 | |
| IgG2a | |
| Protein A | |
| Antibody |