missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Osteopontin Monoclonal Antibody (K52025_9H8), Invitrogen™

Catalog No. MA555760
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555760 50 μL
MA556140 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555760 Supplier Invitrogen™ Supplier No. MA555760
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: DDQSAETHSH KQSRLYKRKA NDESNEHSDV IDSQELSKVS REFHSHEFHS HEDMLVVDPK SKEEDKHLKF

Osteopontin is an arginine-glycine-aspartic acid (RGD)-containing glycoprotein that interacts with integrins and CD44 as major receptors. It is a multifunctional protein involved in bone mineralization, cell adhesion, cell migration, chronic inflammatory disease and transformation. Proteolytic cleavage by thrombin and matrix metalloproteinases close to the integrin-binding Arg-Gly-Asp sequence modulates the function of OPN and its integrin binding properties. Thrombin-cleaved fragments of Osteopontin are overexpressed in malignant glial tumors and provide a molecular niche with survival advantage and provide a novel substrate for plasmin and cathepsin D.

Specifications

Antigen Osteopontin
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K52025_9H8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene SPP1
Gene Accession No. P10451
Gene Alias 2AR; 2b7; 44 kDa bone phosphoprotein; Apl-1; BNSP; Bone sialoprotein 1; Bone sialoprotein 1 (BSPI/BSP1); Bsp; BSPI; Calcium oxalate crystal growth inhibitor protein; CALPHA1 fusion; Early T lymphocyte activation 1 (ETA1); early T-lymphocyte activation 1; early T-lymphocyte activation 1 protein; Eta; ETA-1; HGNC:11255; immunoglobulin alpha 1 heavy chain constant region fusion protein; MGC110940; Minopontin; nephropontin; Op; Opn; Opnl; OSP; osteopontin; osteopontin/immunoglobulin alpha 1 heavy chain constant region fusion protein; osteopontin-like protein; PSEC0156; Ric; Secreted phosphoprotein 1; secreted phosphoprotein 1 (osteopontin bone sialoprotein I early T lymphocyte activation 1); secreted phosphoprotein 1 (osteopontin, bone sialoprotein I, early T-lymphocyte activation 1); Secreted phosphoprotein 1 (SPP1); secreted phosphoprotein 1 variant 6; Sialoprotein (osteopontin); SPP1; SPP-1; SPP1/CALPHA1 fusion; unnamed protein product; urinar; urinary stone protein; Uropontin
Gene Symbols SPP1
Host Species Mouse
Immunogen Recombinant human SPP1 (immunogen range is 231-300).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6696
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less