missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Osteocrin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149813
Description
Osteocrin Polyclonal Antibody for Western Blot, IHC (P)
Hormone that acts as a regulator of dendritic growth in the developing cerebral cortex in response to sensory experience. Induced in the brain following membrane depolarization and inhibits dendritic branching in neurons of the developing cortex.Specifications
| Osteocrin | |
| Polyclonal | |
| Unconjugated | |
| OSTN | |
| MUSCLIN; Ostc; osteocrin; Ostn; Processed Osteocrin | |
| Rabbit | |
| Protein A | |
| RUO | |
| 239790 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P61364 | |
| OSTN | |
| Recombinant Osteocrin (OSTN) (Phe26-Gly130). | |
| 200 μL | |
| Primary | |
| Mouse | |
| Antibody |