missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Osteocalcin Polyclonal Antibody
Guinea Pig Polyclonal Antibody
Supplier: Invitrogen™ PA5149840
Description
Osteocalcin Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Osteocalcin belongs to the osteocalcin/matrix Gla-protein family and constitutes 1-2% of the total bone protein. It binds strongly to apatite and calcium. Gamma-carboxyglutamate residues are formed by vitamin K dependent carboxylation. These residues are essential for the binding of calcium.Specifications
| Osteocalcin | |
| Polyclonal | |
| Unconjugated | |
| BGLAP | |
| Bglap; Bglap1; Bglap2; BGP; Bgpr; Bgpra; bone gamma carboxyglutamate protein; bone gamma carboxyglutamate protein 1; bone gamma-carboxyglutamate (gla) protein; bone gamma-carboxyglutamate (gla) protein (osteocalcin); bone gamma-carboxyglutamate protein; bone gamma-carboxyglutamate protein 2; bone gamma-carboxyglutamate protein osteocalcin; bone gamma-carboxyglutamate; BGLAP; bone gamma-carboxyglutamic acid (Gla) protein (osteocalcin); bone Gla protein; bone gla protein BGP; bone Gla-protein; gamma-carbooxyglutamate (gla) protein; Gamma-carboxyglutamic acid-containing protein; GLA; Gla protein precusor; mOC-A; OC; OCN; OG1; Osteocalcin; PMF1; RP11-54H19.5; unnamed protein product | |
| Guinea Pig | |
| Protein A | |
| RUO | |
| 100340503 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P39056 | |
| BGLAP | |
| Recombinant Osteocalcin (OC) (Lys24-Val100). | |
| 200 μL | |
| Primary | |
| Rabbit | |
| Antibody |