missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Osteocalcin Native Protein

Catalog No. PIRP43153
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP43153 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP43153 Supplier Invitrogen™ Supplier No. RP43153

Native Protein

RP-43153 contains native human Osteocalcin protein. RP-43153 may be used as a positive control in many immunological assays. The product is of human blood/plasma origin. Although the starting material was tested prior to initiation of the manufacturing process, and was found negative or nonreactive for anti-HIV-1/2, HIV-1 antigen(s), HBsAg, STS, anti-HCV, anti-HBcore and anti-HTLV I & II, extreme caution should be used when handling this material as there is a margin of error in all tests. This product is to be used for IN VITRO research purposes only, and is not intended for clinical or diagnostic use!

Osteocalcin belongs to the osteocalcin/matrix Gla-protein family and constitutes 1-2% of the total bone protein. It binds strongly to apatite and calcium. Gamma-carboxyglutamate residues are formed by vitamin K dependent carboxylation. These residues are essential for the binding of calcium.

Specifications

Accession Number P02818
Concentration 0.59 mg/mL
For Use With (Application) Control
Formulation TBS with 2 mM CaCl2 and no preservative; pH 7.4
Gene ID (Entrez) 632
Molecular Weight (g/mol) 5.8 kDa
Name Human Osteocalcin
pH Range 7.4
Quantity 20 μg
Source Human
Gene Alias Bglap; Bglap1; Bglap2; BGP; Bgpr; Bgpra; bone gamma carboxyglutamate protein; bone gamma carboxyglutamate protein 1; bone gamma-carboxyglutamate (gla) protein; bone gamma-carboxyglutamate (gla) protein (osteocalcin); bone gamma-carboxyglutamate protein; bone gamma-carboxyglutamate protein 2; bone gamma-carboxyglutamate protein osteocalcin; bone gamma-carboxyglutamate; BGLAP; bone gamma-carboxyglutamic acid (Gla) protein (osteocalcin); bone Gla protein; bone gla protein BGP; bone Gla-protein; gamma-carbooxyglutamate (gla) protein; Gamma-carboxyglutamic acid-containing protein; GLA; Gla protein precusor; mOC-A; OC; OCN; OG1; Osteocalcin; PMF1; RP11-54H19.5; unnamed protein product
Common Name Osteocalcin
Gene Symbol BGLAP
Conjugate Unconjugated
Species Human
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Form Liquid
Protein Subtype Native
Show More Show Less