missing translation for 'onlineSavingsMsg'
Learn More

Osmium(VIII) oxide, 99.8% (metals basis), Os 74.4% min

Catalog No. p-4782035
Change view
Click to view available options
Quantity:
0.5 g
1 g
10 x 1 g
Catalog No. Quantity
AA1210302 0.5 g
AA1210303 1 g
AA1210348 10 x 1 g
3 options

Catalog No. AA1210302

Supplier: Thermo Scientific Chemicals 01210302

Only null left

CAS: 20816-12-0 | O4Os | 254.23 g/mol

Oxidation, catalyst in Sharpless dihydroxylations Osmium(VIII) oxide is used in the Sharpless oxyamination to give vicinal amino-alcohols. It is used to oxidize alkenes to the vicinal diols. It acts as a staining agent used in transmission electron microscopy to provide contrast to the image and as a stain for lipids in optical microscopy. As a polymer stain, it is also used to stain selectively copolymers especially styrene-butadiene block copolymers. Further, it is involved in the synthesis of Isosteviol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 20816-12-0
Molecular Formula O4Os
Molecular Weight (g/mol) 254.23
MDL Number MFCD00011150
InChI Key VUVGYHUDAICLFK-UHFFFAOYSA-N
Synonym osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
PubChem CID 30318
IUPAC Name tetraoxoosmium
SMILES O=[Os](=O)(=O)=O

Specifications

Boiling Point 130°C
Melting Point 40.6°C
Physical Form Crystalline
Quantity 0.5 g
Assay Percent Range 99.8% (metals basis)
Solubility Information Soluble in chloroform,alcohol and ethers.Soluble in water,organic solvents,benzene,alcohol,ether,ammonium hydroxide,phosphorus oxychloride and carbon tetrachloride.
Formula Weight 254.1
Odor Like chlorine
Appearance Crystalline
Vapor Pressure 10 hPa (8mm Hg) at 20°C
Chemical Name or Material Osmium(VIII) oxide
Show More Show Less
Hazard Category H300+H310+H330-H314-H335
Hazard Statement Keep container tightly closed and in a well-ventilated place. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation DOT Class: 6.1, Packaging Group: I
EINECSNumber 244-058-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only