missing translation for 'onlineSavingsMsg'
Learn More

Osmium(VIII) oxide, 4% aq. soln.

Catalog No. p-4407677
Change view
Click to view available options
Quantity:
2 mL
10 mL
Catalog No. Quantity
AA453859W 2 mL
AA45385AB 10 mL
2 options

Catalog No. AA453859W

Supplier: Thermo Scientific Chemicals 0453859W

Only null left

CAS: 20816-12-0 | O4Os | 254.23 g/mol

Osmium(VIII) oxide is widely used as an oxidant in organic synthesis to oxidize alkenes to vicinal diols as well as in the sharpless oxyamination to give vicinal amino-alcohols. It is involved in the oxidative cleavage of alkenes (Lemieux-Johnson oxidation) in association with sodium periodate. It finds application in transmission electron microscopy (TEM) as a staining agent. It is involved in the promotion of oxidative cleavage of graphene layers in carbon nanotubes and as an intermediate in osmium ore refining. As a lipid stain, it is useful in scanning electron microscopy (SEM) as an alternative to sputter coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20816-12-0
Molecular Formula O4Os
Molecular Weight (g/mol) 254.23
MDL Number MFCD00011150
InChI Key VUVGYHUDAICLFK-UHFFFAOYSA-N
Synonym osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
PubChem CID 30318
IUPAC Name tetraoxoosmium
SMILES O=[Os](=O)(=O)=O

Specifications

Physical Form Liquid
Quantity 2 mL
Solubility Information Soluble in alcohol,ether,chloroform,benzene,ammonium hydroxide,phosphorus oxychloride and carbon tetrachloride
Formula Weight 254.1
Sensitivity Air Sensitive
Vapor Pressure 17mm Hg at 20°C
Chemical Name or Material Osmium(VIII) oxide
Hazard Category H310-H315-H318-H332-H334
Precautionary Statement P261-P262-P264b-P270-P271-P280-P285-P302+P350-P304+P340-P305+P351+P338-P310-P332+P313-P342+P311-P361-P363-P501c
DOTInformation DOT Class: 6.1; Packaging Group: II
EINECSNumber 244-058-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only