Learn More
Osmium(VIII) oxide, 2% aq. soln.
CAS: 20816-12-0 | O4Os | 254.23 g/mol
$162.93 - $250.67
Chemical Identifiers
| CAS | 20816-12-0 |
|---|---|
| Molecular Formula | O4Os |
| Molecular Weight (g/mol) | 254.23 |
| MDL Number | MFCD00011150 |
| InChI Key | VUVGYHUDAICLFK-UHFFFAOYSA-N |
| Synonym | osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4 |
| PubChem CID | 30318 |
| IUPAC Name | tetraoxoosmium |
| SMILES | O=[Os](=O)(=O)=O |
Description
Osmium(VIII) oxide is widely used as an oxidant in organic synthesis to oxidize alkenes to vicinal diols as well as in the sharpless oxyamination to give vicinal amino-alcohols. It is involved in the oxidative cleavage of alkenes (Lemieux-Johnson oxidation) in association with sodium periodate. It finds application in transmission electron microscopy (TEM) as a staining agent. It is involved in the promotion of oxidative cleavage of graphene layers in carbon nanotubes and as an intermediate in osmium ore refining. As a lipid stain, it is useful in scanning electron microscopy (SEM) as an alternative to sputter coating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 20816-12-0 | |
| 254.23 | |
| VUVGYHUDAICLFK-UHFFFAOYSA-N | |
| 30318 | |
| O=[Os](=O)(=O)=O |
| O4Os | |
| MFCD00011150 | |
| osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4 | |
| tetraoxoosmium |
Specifications
| 20816-12-0 | |
| Liquid | |
| O4Os | |
| UN3287 | |
| osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4 | |
| VUVGYHUDAICLFK-UHFFFAOYSA-N | |
| tetraoxoosmium | |
| 30318 | |
| 2% aq. soln. | |
| Air Sensitive |
| Colorless to Yellow | |
| 2 mL | |
| MFCD00011150 | |
| 14,6893 | |
| Soluble in alcohol,ether,chloroform,benzene,ammonium hydroxide,phosphorus oxychloride and carbon tetrachloride | |
| O=[Os](=O)(=O)=O | |
| 254.23 | |
| 254.1 | |
| Chlorine-like | |
| Osmium (VIII) oxide |
Safety and Handling
GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P361-P363-P501c
H311-H315-H319-H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC LIQUID, INORGANIC, N.O.S.
EINECSNumber : 244-058-7
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only