missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OSGIN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149957
Description
OSGIN1 Polyclonal Antibody for Western Blot, ICC/IF, IHC
OSGIN1 regulates the differentiation and proliferation of normal cells through the regulation of cell death.Specifications
| OSGIN1 | |
| Polyclonal | |
| Unconjugated | |
| Osgin1 | |
| 1700012B18Rik; BDGI; BMSC-derived growth inhibitor; Bone marrow stromal cell-derived growth inhibitor; huOKL38; LOW QUALITY PROTEIN: oxidative stress-induced growth inhibitor 1; Okl38; OSGIN1; Ovary, kidney and liver protein 38; oxidative stress induced growth inhibitor 1; oxidative stress-induced growth inhibitor 1; pregnancy-induced growth inhibitor; pregnancy-induced growth inhibitor OKL38; Unknown (protein for MGC:137469) | |
| Rabbit | |
| Protein A | |
| RUO | |
| 29948 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9UJX0 | |
| Osgin1 | |
| Recombinant Oxidative Stress Induced Growth Inhibitor 1 (OSGIN1) (Gly25-Val281). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody |