missing translation for 'onlineSavingsMsg'
Learn More

Oseltamivir phosphate, 98%, Thermo Scientific Chemicals

$160.45 - $160.45

Chemical Identifiers

CAS 204255-11-8
Molecular Formula C16H28N2O4·H3PO4
Molecular Weight (g/mol) 410.4
InChI Key PGZUMBJQJWIWGJ-ONAKXNSWSA-N
Synonym oseltamivir phosphate, tamiflu, oseltamir phosphate, unii-4a3o49ngez, 4a3o49ngez, ethyl 3r,4r,5s-4-acetamido-5-amino-3-pentan-3-yloxy cyclohex-1-ene-1-carboxylate phosphate, ethyl 3r,4r,5s-4-acetamido-5-amino-3-1-ethylpropoxy-1-cyclohexene-1-carboxylate phosphate 1:1, osteltamivir phosphate, 3r-3alpha,4beta,5alpha-ethyl 4-acetylamino-5-amino-3-1-ethylpropoxy-1-cyclohexene-1-carboxylate phosphate 1:1
PubChem CID 78000
ChEBI CHEBI:7799
IUPAC Name ethyl (3R,4R,5S)-4-acetamido-5-amino-3-pentan-3-yloxycyclohexene-1-carboxylate;phosphoric acid
SMILES CCC(CC)OC1C=C(CC(C1NC(=O)C)N)C(=O)OCC.OP(=O)(O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC461170010
View Documents
Thermo Scientific Chemicals
461170010
1 g
Each for $160.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

204255-11-8
410.4
oseltamivir phosphate, tamiflu, oseltamir phosphate, unii-4a3o49ngez, 4a3o49ngez, ethyl 3r,4r,5s-4-acetamido-5-amino-3-pentan-3-yloxy cyclohex-1-ene-1-carboxylate phosphate, ethyl 3r,4r,5s-4-acetamido-5-amino-3-1-ethylpropoxy-1-cyclohexene-1-carboxylate phosphate 1:1, osteltamivir phosphate, 3r-3alpha,4beta,5alpha-ethyl 4-acetylamino-5-amino-3-1-ethylpropoxy-1-cyclohexene-1-carboxylate phosphate 1:1
CHEBI:7799
CCC(CC)OC1C=C(CC(C1NC(=O)C)N)C(=O)OCC.OP(=O)(O)O
C16H28N2O4·H3PO4
PGZUMBJQJWIWGJ-ONAKXNSWSA-N
78000
ethyl (3R,4R,5S)-4-acetamido-5-amino-3-pentan-3-yloxycyclohexene-1-carboxylate;phosphoric acid

Specifications

204255-11-8
Authentic
C16H28N2O4·H3PO4
oseltamivir phosphate, tamiflu, oseltamir phosphate, unii-4a3o49ngez, 4a3o49ngez, ethyl 3r,4r,5s-4-acetamido-5-amino-3-pentan-3-yloxy cyclohex-1-ene-1-carboxylate phosphate, ethyl 3r,4r,5s-4-acetamido-5-amino-3-1-ethylpropoxy-1-cyclohexene-1-carboxylate phosphate 1:1, osteltamivir phosphate, 3r-3alpha,4beta,5alpha-ethyl 4-acetylamino-5-amino-3-1-ethylpropoxy-1-cyclohexene-1-carboxylate phosphate 1:1
CCC(CC)OC1C=C(CC(C1NC(=O)C)N)C(=O)OCC.OP(=O)(O)O
1 g
78000
410.4
Oseltamivir phosphate
White to Off-White
97.5% min. (HPLC)
Glass Bottle
PGZUMBJQJWIWGJ-ONAKXNSWSA-N
ethyl (3R,4R,5S)-4-acetamido-5-amino-3-pentan-3-yloxycyclohexene-1-carboxylate;phosphoric acid
410.4
CHEBI:7799
98%
Videos
Safety and Handling

Safety and Handling

Causes serious eye irritation, Harmful if swallowed, May cause respiratory irritation, Causes skin irritation

Serious eye damage/eye irritation (category 2), Acute toxicity (category 4), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)

SDS
Documents

Documents

RUO – Research Use Only