missing translation for 'onlineSavingsMsg'
Learn More

OSBPL11 Antibody, ProSci

Catalog No. 16592442
Change view
Click to view available options
Quantity:
0.4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16592442 0.4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16592442 Supplier ProSci Llc Supplier No. 599630400

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

OSBPL11 Polyclonal Antibody specifically detects RNF41 in Human Samples. It is validated for Western Blot.

Specifications

Antigen OSBPL11
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution For WB starting dilution is: 1:1000
Formulation Supplied in PBS with 0.09% (W/V) sodium azide.
Gene Accession No. Q9BXB4
Gene Alias Oxysterol-binding protein-related protein 11, ORP-11, OSBP-related protein 11, OSBPL11, ORP11, OSBP12
Host Species Rabbit
Purification Method This antibody is purified through a protein A column, followed by peptide affinity purification.
Quantity 0.4 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114885
Target Species Human
Content And Storage Store at 4°C for three months and -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Product Type Antibody
Form Liquid
Isotype Ig
Research Category Obesity, Signal Transduction
Show More Show Less