missing translation for 'onlineSavingsMsg'
Learn More

Orphenadrine Citrate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014234
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014234 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014234 Supplier Merck Emd Millipore Supplier No. PHR17771G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Orphenadrine Citrate is an analog of diphenhydramine antihistamine drug. It operates as a centrally acting skeletal muscle relaxant and effectively serves to relieve painful skeletal muscle spasms by working as an analgesics.

Specifications

CAS 4682-36-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C18H23 NO · C6H8O7
MDL Number MFCD00079197
Synonym Orphenadrine citrate salt; beta-Dimethylaminoethyl 2-methylbenzhydryl ether citrate salt; o-Methyldiphenhydramine citrate salt
Type Certified Reference Material
Quantity 1 g
Formula Weight 461.50
Grade Certified Reference Material