missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ORP7 Polyclonal Antibody

Description
Immunogen sequence: DQLRDMHQGS ELSRMGVSEA STGQRRLHSL STSSDTTADS FSSLNPEEQE ALYMKGRELT PQLSQTSILS LADSHTEF Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 88%.
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Two transcript variants encoding the same isoform have been identified.
Specifications
Specifications
| Antigen | ORP7 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | OSBPL7 |
| Gene Accession No. | Q9BZF2 |
| Gene Alias | ORP7; ORP-7; OSBPL7; OSBP-related protein 7; oxysterol binding protein like 7; oxysterol binding protein-like 7; oxysterol-binding protein-related protein 7 |
| Gene Symbols | OSBPL7 |
| Show More |