missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ORP1 Recombinant Rabbit Monoclonal Antibody (23GB4065)
GREENER_CHOICE

Catalog No. MA553684
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553684 100 μL
MA553685 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553684 Supplier Invitrogen™ Supplier No. MA553684
Only null left

Rabbit Recombinant Monoclonal Antibody

ORP1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene encodes a member of the oxysterol-binding protein family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain, although some members contain only the sterol-binding domain. Transcript variants derived from alternative promoter usage and/or alternative splicing exist; they encode different isoforms.

Specifications

Antigen ORP1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4065
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OSBPL1A
Gene Accession No. Q8K4M9, Q91XL9, Q9BXW6
Gene Alias DCDC4A; FLJ10217; FLJ50293; FLJ55454; FLJ79410; G430090F17Rik; Gm753; ORP1; ORP-1; Orp1a; Orp1l; OSBP8; OSBPL1; OSBPL1A; OSBPL1B; OSBP-related protein 1; oxysterol binding protein like 1A; oxysterol binding protein-like 1A; oxysterol binding protein-like 1B; oxysterol-binding protein-related protein 1; oxysterol-binding protein-related protein 1 variant 1; oxysterol-binding protein-related protein 1 variant 2
Gene Symbols OSBPL1A
Host Species Rabbit
Immunogen A synthesized peptide derived from human ORP1 (920-950AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114876, 259221, 64291
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less