missing translation for 'onlineSavingsMsg'
Learn More

ORP test solution, 200-275 mV

Catalog No. AA42131AK
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42131AK 250 mL
AA42131AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42131AK Supplier Thermo Scientific Chemicals Supplier No. 042131AK

Test solution for ORP electrodes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material ORP test solution
Packaging Plastic-coated glass bottle
Physical Form Liquid
MDL Number MFCD01867801
Quantity 250 mL

RUO – Research Use Only